Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
WFS1
Entrez Gene ID
101112732
Gene Name
wolframin ER transmembrane glycoprotein
Gene Aliases
-
Chromosome Location
Chr.6: 113270691 - 113302426 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971171.1 XP_027826972.1 6 - 7 849 54

View Full Product Details