Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APBA1
Entrez Gene ID
101105978
Gene Name
amyloid beta precursor protein binding family A member 1
Gene Aliases
-
Chromosome Location
Chr.2: 72111647 - 72346762 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093185.2 XP_014948671.2 8 - 9 1970 65

View Full Product Details