Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAVER2
Entrez Gene ID
101110463
Gene Name
ribonucleoprotein, PTB binding 2
Gene Aliases
-
Chromosome Location
Chr.1: 42200184 - 42362326 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012167788.3 XP_012023178.2 10 - 11 2498 65

View Full Product Details