Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD1
Entrez Gene ID
101106630
Gene Name
BTB domain containing 1
Gene Aliases
-
Chromosome Location
Chr.18: 20918029 - 20965342 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957101.1 XP_027812902.1 4 - 5 999 71
XM_027957102.1 XP_027812903.1 4 - 5 921 71
XM_027957103.1 XP_027812904.1 4 - 5 921 71

View Full Product Details