Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEND2
Entrez Gene ID
101103875
Gene Name
BEN domain containing 2
Gene Aliases
-
Chromosome Location
Chr.X: 17215905 - 17271868 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963216.1 XP_027819017.1 5 - 6 890 71

View Full Product Details