Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPH4
Entrez Gene ID
101121939
Gene Name
junctophilin 4
Gene Aliases
-
Chromosome Location
Chr.7: 22685092 - 22693983 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971617.1 XP_027827418.1 2 - 3 1232 62

View Full Product Details