Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TGFB2
Entrez Gene ID
554322
Gene Name
transforming growth factor beta 2
Gene Aliases
-
Chromosome Location
Chr.12: 23881886 - 23975387 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013601.3 XP_004013650.1 5 - 6 1583 59
XM_004013602.3 XP_004013651.1 6 - 7 1669 59
GenBank mRNA AY656797.1 - 5 - 6 911 59

View Full Product Details