Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEG1
Entrez Gene ID
101121656
Gene Name
heart development protein with EGF like domains 1
Gene Aliases
-
Chromosome Location
Chr.1: 207495674 - 207579687 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027956598.1 XP_027812399.1 12 - 13 3989 55
XM_027956605.1 XP_027812406.1 12 - 13 3985 55

View Full Product Details