Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPATCH3
Entrez Gene ID
101109595
Gene Name
G-patch domain containing 3
Gene Aliases
-
Chromosome Location
Chr.2: 255837092 - 255844057 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005088.4 XP_004005137.2 6 - 7 1371 60

View Full Product Details