Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCAKD
Entrez Gene ID
101117519
Gene Name
dephospho-CoA kinase domain containing
Gene Aliases
-
Chromosome Location
Chr.11: 18317026 - 18344851 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012993.4 XP_004013042.1 4 - 5 601 73
XM_012186223.3 XP_012041613.1 5 - 6 816 73

View Full Product Details