Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSX1
Entrez Gene ID
101106277
Gene Name
visual system homeobox 1
Gene Aliases
-
Chromosome Location
Chr.13: 44179100 - 44187251 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976427.1 XP_027832228.1 3 - 4 627 59

View Full Product Details