Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PBX4
Entrez Gene ID
101104316
Gene Name
PBX homeobox 4
Gene Aliases
-
Chromosome Location
Chr.5: 3753307 - 3816009 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969340.1 XP_027825141.1 6 - 7 1009 93

View Full Product Details