Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LCA5
Entrez Gene ID
101113154
Gene Name
lebercilin LCA5
Gene Aliases
-
Chromosome Location
Chr.8: 7315894 - 7371899 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972300.1 XP_027828101.1 3 - 4 981 102

View Full Product Details