Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAS1R2
Entrez Gene ID
101106153
Gene Name
taste 1 receptor member 2
Gene Aliases
-
Chromosome Location
Chr.2: 263302592 - 263319549 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964000.1 XP_027819801.1 1 - 2 174 67

View Full Product Details