Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCBE1
Entrez Gene ID
101117553
Gene Name
collagen and calcium binding EGF domains 1
Gene Aliases
-
Chromosome Location
Chr.23: 64461830 - 64691378 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015103800.2 XP_014959286.2 5 - 6 726 60

View Full Product Details