Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPAT3
Entrez Gene ID
101119394
Gene Name
glycerol-3-phosphate acyltransferase 3
Gene Aliases
AGPAT9
Chromosome Location
Chr.6: 108595873 - 108658580 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012180288.3 XP_012035678.1 10 - 11 1366 101
XM_027971150.1 XP_027826951.1 10 - 11 1441 101

View Full Product Details