Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LENG1
Entrez Gene ID
101102751
Gene Name
leukocyte receptor cluster member 1
Gene Aliases
-
Chromosome Location
Chr.14: 68155291 - 68158711 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015435.4 XP_004015484.2 3 - 4 648 58

View Full Product Details