Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCL1
Entrez Gene ID
554329
Gene Name
MCL1 apoptosis regulator, BCL2 family member
Gene Aliases
Mcl-1
Chromosome Location
Chr.1: 106341279 - 106346576 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015092234.2 XP_014947720.2 2 - 3 1631 87
GenBank mRNA AF144097.1 - 1 - 2 196 87

View Full Product Details