Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
NCS1
Entrez Gene ID
101102136
Gene Name
neuronal calcium sensor 1
Gene Aliases
-
Chromosome Location
Chr.3: 6508338 - 6564752 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966284.1 XP_027822085.1 1 - 2 112 80

View Full Product Details