Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NIPAL1
Entrez Gene ID
101101985
Gene Name
NIPA like domain containing 1
Gene Aliases
-
Chromosome Location
Chr.6: 73201190 - 73224851 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027970980.1 XP_027826781.1 2 - 3 403 60

View Full Product Details