Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLF10
Entrez Gene ID
101104073
Gene Name
Kruppel like factor 10
Gene Aliases
-
Chromosome Location
Chr.9: 82600100 - 82606153 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027973155.1 XP_027828956.1 2 - 3 372 103

View Full Product Details