Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK7
Entrez Gene ID
101108703
Gene Name
potassium two pore domain channel subfamily K member 7
Gene Aliases
-
Chromosome Location
Chr.21: 45568994 - 45571835 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019672.4 XP_004019721.1 2 - 3 771 90

View Full Product Details