Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ODF3L1
Entrez Gene ID
101104861
Gene Name
outer dense fiber of sperm tails 3 like 1
Gene Aliases
-
Chromosome Location
Chr.18: 31015796 - 31019499 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017835.4 XP_004017884.1 2 - 3 319 97

View Full Product Details