Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DBNDD2
Entrez Gene ID
101109472
Gene Name
dysbindin domain containing 2
Gene Aliases
-
Chromosome Location
Chr.13: 76961150 - 76966680 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012189241.3 XP_012044631.2 2 - 3 498 89
XM_012189242.3 XP_012044632.2 2 - 3 202 89
XM_012189244.3 XP_012044634.2 2 - 3 574 89
XM_027977239.1 XP_027833040.1 2 - 3 1013 89
XM_027977240.1 XP_027833041.1 2 - 3 225 89

View Full Product Details