Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COQ6
Entrez Gene ID
101105664
Gene Name
coenzyme Q6, monooxygenase
Gene Aliases
-
Chromosome Location
Chr.7: 89096103 - 89110060 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011068.4 XP_004011117.2 7 - 8 843 58
XM_027972006.1 XP_027827807.1 7 - 8 832 58
XM_027972007.1 XP_027827808.1 7 - 8 813 58
XM_027972008.1 XP_027827809.1 7 - 8 813 58

View Full Product Details