Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPOCD1
Entrez Gene ID
101106499
Gene Name
SPOC domain containing 1
Gene Aliases
-
Chromosome Location
Chr.2: 251268937 - 251301891 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965366.1 XP_027821167.1 7 - 8 2308 73

View Full Product Details