Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM81A
Entrez Gene ID
101115214
Gene Name
family with sequence similarity 81 member A
Gene Aliases
-
Chromosome Location
Chr.7: 52160907 - 52220040 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971781.1 XP_027827582.1 8 - 9 1033 81
XM_027971782.1 XP_027827583.1 7 - 8 1027 81
XM_027971783.1 XP_027827584.1 8 - 9 1062 81
XM_027971784.1 XP_027827585.1 7 - 8 1056 81

View Full Product Details