Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THG1L
Entrez Gene ID
101113674
Gene Name
tRNA-histidine guanylyltransferase 1 like
Gene Aliases
-
Chromosome Location
Chr.5: 72268631 - 72275635 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004009022.4 XP_004009071.3 2 - 3 377 71
XM_012178866.3 XP_012034256.2 2 - 3 349 71

View Full Product Details