Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCR7
Entrez Gene ID
101109462
Gene Name
C-C motif chemokine receptor 7
Gene Aliases
-
Chromosome Location
Chr.11: 22394564 - 22408016 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012868.4 XP_004012917.1 2 - 3 98 84

View Full Product Details