Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A1BG
Entrez Gene ID
101119665
Gene Name
alpha-1-B glycoprotein
Gene Aliases
-
Chromosome Location
Chr.14: 70903705 - 70908207 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978951.1 XP_027834752.1 3 - 4 697 75

View Full Product Details