Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
RUNX3
Entrez Gene ID
101119625
Gene Name
RUNX family transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.2: 257536339 - 257602751 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965500.1 XP_027821301.1 2 - 3 475 87

View Full Product Details