Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMAIP1
Entrez Gene ID
101102359
Gene Name
phorbol-12-myristate-13-acetate-induced protein 1
Gene Aliases
-
Chromosome Location
Chr.23: 64825502 - 64829813 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020571.4 XP_004020620.1 1 - 2 235 99

View Full Product Details