Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THEM4
Entrez Gene ID
101119606
Gene Name
thioesterase superfamily member 4
Gene Aliases
-
Chromosome Location
Chr.1: 107574249 - 107625493 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975113.1 XP_027830914.1 2 - 3 367 107

View Full Product Details