Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCN10
Entrez Gene ID
101120313
Gene Name
lipocalin 10
Gene Aliases
-
Chromosome Location
Chr.3: 1140924 - 1143187 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968306.1 XP_027824107.1 4 - 5 646 90

View Full Product Details