Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEMGN
Entrez Gene ID
101103212
Gene Name
hemogen
Gene Aliases
-
Chromosome Location
Chr.2: 52430486 - 52441934 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963903.1 XP_027819704.1 2 - 3 250 66

View Full Product Details