Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZDHHC22
Entrez Gene ID
101112985
Gene Name
zinc finger DHHC-type palmitoyltransferase 22
Gene Aliases
-
Chromosome Location
Chr.7: 92334714 - 92343628 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015097204.2 XP_014952690.2 2 - 2 1448 107

View Full Product Details