Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPN1MW
Entrez Gene ID
100008674
Gene Name
opsin 1, medium wave sensitive
Gene Aliases
GCP, OPN1MW2
Chromosome Location
Chr.Unknown: 293885 - 304206 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322264.1 NP_001309193.1 - 599 63
GenBank mRNA AF031526.1 - - 435 63
DN891010.1 - - 599 63

View Full Product Details