Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRD
Entrez Gene ID
100008933
Gene Name
chordin
Gene Aliases
CHD
Chromosome Location
Chr.14: 81327039 - 81334111 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008266589.1 XP_008264811.1 10 - 11 1979 142
XM_017346951.1 XP_017202440.1 10 - 11 1979 142
XM_017346952.1 XP_017202441.1 - 1979 142
GenBank mRNA AY575210.1 - 1 - 2 88 142

View Full Product Details