Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LPH
- Entrez Gene ID
-
100009256
- Gene Name
- lactase phlorizin hydrolase
- Gene Aliases
-
LCT
- Chromosome Location
-
Chr.7: 73309071 - 73368266 on Build OryCun2.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001101689.1 |
NP_001095159.1 |
16 - 17 |
5604 |
131 |
GenBank mRNA |
X07995.1 |
- |
16 - 17 |
5604 |
131 |
- Gene Symbol
- LCT
- Entrez Gene ID
-
100352497
- Gene Name
- lactase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 73090090 - 73138978 on Build OryCun2.0
- Gene Symbol
- LOC100353264
- Entrez Gene ID
-
100353264
- Gene Name
- lactase-phlorizin hydrolase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 73183796 - 73266579 on Build OryCun2.0
View Full Product Details