Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNB1
Entrez Gene ID
100008779
Gene Name
potassium voltage-gated channel subfamily B member 1
Gene Aliases
Shab
Chromosome Location
Chr.Unknown: 3295976 - 3390629 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082087.1 NP_001075556.1 - 2445 111
GenBank mRNA AF266507.1 - - 2445 111

View Full Product Details