Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GATA4
Entrez Gene ID
100008858
Gene Name
GATA binding protein 4
Gene Aliases
GATA-4
Chromosome Location
Chr.2: 38220947 - 38299571 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002709438.3 XP_002709484.1 3 - 4 1593 84
XM_008248879.2 XP_008247101.1 3 - 4 1597 84
XM_008248888.2 XP_008247110.1 3 - 4 1316 84
XM_017337712.1 XP_017193201.1 3 - 4 1328 84
GenBank mRNA AJ291310.1 - 1 - 2 157 84

View Full Product Details