Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLA-DR-ALPHA
Entrez Gene ID
100328929
Gene Name
major histocompatibility complex, class II, DR alpha
Gene Aliases
-
Chromosome Location
Chr.12: 23070319 - 23074942 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171118.1 NP_001164589.1 2 - 3 393 87

View Full Product Details