Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMOD4
Entrez Gene ID
100328666
Gene Name
tropomodulin 4
Gene Aliases
-
Chromosome Location
Chr.13: 41017697 - 41022304 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171299.1 NP_001164770.1 4 - 5 487 64
XM_008264027.2 XP_008262249.1 6 - 7 793 64
XM_008264029.2 XP_008262251.1 5 - 6 666 64
XM_017345510.1 XP_017200999.1 5 - 6 673 64
XM_017345511.1 XP_017201000.1 5 - 6 635 64
XM_017345512.1 XP_017201001.1 4 - 5 507 64

View Full Product Details