Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM6
Entrez Gene ID
100328879
Gene Name
tripartite motif containing 6
Gene Aliases
-
Chromosome Location
Chr.1: 146617976 - 146629510 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171472.1 NP_001164943.1 1 - 2 420 94

View Full Product Details