Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
OR52N4
Entrez Gene ID
100328885
Gene Name
olfactory receptor, family 52, subfamily N, member 4
Gene Aliases
-
Chromosome Location
Chr.1: 146760457 - 146761376 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171476.1 NP_001164947.1 1 - 1 51 78

View Full Product Details