Gene Transcripts

Gene Symbol
Slc4a2
Entrez Gene ID
24780
Gene Name
solute carrier family 4 (anion exchanger), member 2
Gene Aliases
Ae2, Aep2
Chromosome Location
Chr. 4: 7264678 - 7282734 on Build Rnor_6.0
UniGene ID
Rn.9860
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017048.2 NP_058744.1 20 3277
XM_006235874.2 XP_006235936.1 20 3734
XM_006235875.2 XP_006235937.1 20 3455
XM_006235876.2 XP_006235938.1 20 3517
XM_006235877.2 XP_006235939.1 19 3660
XM_006235878.2 XP_006235940.1 19 3579
GenBank mRNA J05166.1 - 20 3281

View Full Product Details