Gene Transcripts

Gene Symbol
Tfr2
Entrez Gene ID
288562
Gene Name
transferrin receptor 2
Gene Aliases
Trfr2
Chromosome Location
Chr. 12: 22177382 - 22194333 on Build Rnor_6.0
UniGene ID
Rn.59926
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105916.1 NP_001099386.1 20 2567
XM_006249116.2 XP_006249178.1 19 2519
XM_006249117.1 XP_006249179.1 20 2535
XM_006249118.1 XP_006249180.1 20 2538
XM_008769125.1 XP_008767347.1 19 2648
XM_008769126.1 XP_008767348.1 19 2478
XM_008769127.1 XP_008767349.1 18 2291
XM_008769128.1 XP_008767350.1 19 2571
GenBank mRNA BC166451.1 - 19 2632

View Full Product Details