Gene Transcripts

Gene Symbol
Ccdc61
Entrez Gene ID
292680
Gene Name
coiled-coil domain containing 61
Gene Aliases
RGD1307390
Chromosome Location
Chr. 1: 79796557 - 79816536 on Build Rnor_6.0
UniGene ID
Rn.34772
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106229.1 NP_001099699.1 6 1078
GenBank mRNA BC127532.1 - 6 980

View Full Product Details