Gene Transcripts

Gene Symbol
Olr79
Entrez Gene ID
293222
Gene Name
olfactory receptor 79
Gene Aliases
MOR6-1
Chromosome Location
Chr. 1: 168244888 - 168245829 on Build Rnor_6.0
UniGene ID
Rn.141907
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002270.1 NP_001002270.1 1 681

View Full Product Details