Gene Transcripts

Gene Symbol
Tex10
Entrez Gene ID
298065
Gene Name
testis expressed 10
Gene Aliases
Chromosome Location
Chr. 5: 64179247 - 64225844 on Build Rnor_6.0
UniGene ID
Rn.20483
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106653.1 NP_001100123.1 8, 9 2153
XM_006238034.2 XP_006238096.1 7, 8 3179

View Full Product Details